tri-Sodium citrate dihydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Sodium Citrate Dihydrate, Sodium citrate tribasic dihydrate, Sodium Citrate, Dihydrate, Sodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate, Citric acid trisodium salt dihydrate
CAS Number:6132-04-3
:
Formula:Na₃C₆H₅O₇·2H₂O
MDL Number:MFCD00150031
Density:1.76 g/cm³ (20 °C)
Molecular Weight:294.1 g/mol
Melting Pt:150
Merck Index:13,08675
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:200-675-3
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together