Potassium permanganate 0.0025 mol/l (0.0125 N) in aqueous solution
Supplier: VWR Collection
Certificates
About this item
Synonyms: Permanganic acid potassium salt
CAS Number:7722-64-7
:
Formula:KMnO₄
MDL Number:MFCD00011364
Molecular Weight:158.03 g/mol
ADR:9 III
Merck Index:12,07824
EINECS:231-760-3
UN:3082
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together