Sodium silicate in aqueous solution, TECHNICAL
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Silicic acid sodium salt, Sodium silicate
CAS Number:1344-09-8
:
Formula:Na₂O(SiO₂)x·xH₂O
MDL Number:MFCD00003492
Density:1,34 g/cm³
ADR:8 III
Merck Index:13,08751
EINECS:215-687-4
UN:3266
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Silica:25.6 - 27.6 %
- Sodium oxide:7.5 - 8.5 %
Specification Test Results
Related Information
Certificates
Search by Lot Number2,5L Plastic bottle for liquids DIN 45 UN X (ZPACK13962500) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Packaging Information
spec_VWRC28079.320 spec_VWRC28079.320 spec_VWRC28079.320 spec_VWRC28079.363 spec_VWRC28079.363
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Frequently Bought Together