Ethanol 90% 90% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Ethyl alcohol
CAS Number:64-17-5
:
Formula:C₂H₅OH
MDL Number:MFCD00003568
Boiling Pt:78.3
Molecular Weight:46.07 g/mol
Melting Pt:-117
Flash Pt:12
ADR:3 II
Merck Index:12,03806
EINECS:200-578-6
UN:1170
:GHS02
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:Min. 89.5 %
- Density (20/20):0.827 - 0.833 kg/l
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC83811.410_T
Product specification
VWR Chemicals Big Packs and Bulks
Product Literature
Frequently Bought Together