Order Entry
United Kingdom
ContactUsLinkComponent
Light distillate suitable for use in the testing of petroleum products by IP and ASTM methods
undefined
Light distillate suitable for use in the testing of petroleum products by IP and ASTM methods
  • Environmentally Preferable:
  • Aromatics (W/V):
    Max. 1.00 %
  • Distillation range (98 %):
    185 - 280°C
Frequently Bought Together