Biuret reagent solution for protein (total) in serum or plasma
Catalog # 220947S
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic container
- Pk:5 L
- Seq:5190
- Environmentally Preferable:
Specifications
Specification Test Results
- Suitability:Passes test
Specification Test Results
About this item
:
UN:3266
:GHS07
Storage Temperature:Ambient
:108-19-0
ADR:8 II