Order Entry
United Kingdom
ContactUsLinkComponent
Microscope diamond slide writers
  SIMPM795-1
undefined
Microscope diamond slide writers
  SIMPM795-1
  • Description:
    Diamond styluses microscope slide labellers
  • Environmentally Preferable:
  • Pk:
    6
The microscope diamond slide writer is compatible with a wide variety of slide labellers.
  • Consistent even coating of diamond dust
  • Stainless steel shaft
This diamond stylus is perfectly suited for the Thermo Scientific PrintMate and MicroWriter, RA Lamb, and TBS Shurmark microscope slide labellers. Manufactured with a consistent even coating of diamond dust.
The 2,35 mm shaft is made from stainless steel.