Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Sera-Xtracta genomic DNA kit

Sera-Xtracta genomic DNA kit

Sera-Xtracta Genomic DNA Kit, 1 KIT
Catalog # 29-4291-40
Supplier:   Cytiva
Sera-Xtracta genomic DNA kit
Catalog # 29-4291-40
Supplier:  Cytiva
CAS Number:  
Sera-Xtracta genomic DNA kit
Sera-Xtracta genomic DNA kit

Sera-Xtracta Genomic DNA Kit, 1 KIT

CAS Number:  
Price
£382.00
Log in for availability
1 * 1 KIT
Product Details & Documents

Sera-Xtracta genomic DNA kit offers magnetic bead-based extraction and purification of genomic DNA from whole blood.

  • High recovery rates: 4 to 8 μg from 200 μl of whole blood
  • High purity genomic DNA: Purity ratio (A260/A280) greater than 1,7
  • Simplified protocol minimises the co-purification of RNA removing the need for RNase treatment in most applications
  • Compatible with molecular biology techniques, including next generation sequencing (NGS), cloning, restriction enzyme digestion, PCR amplification and genotyping applications

The kit yields genomic DNA with the high purity and quality required by today’s molecular biology techniques.

The kit uses chaotropic agents to extract DNA from blood cells, denature protein components and promote the selective binding of DNA to the silica-coated magnetic beads. Proteinase K is the protease of choice to digest protein from samples, because it is active even when enzyme inhibitors such as EDTA and detergents are present in samples. Denatured contaminants are easily removed by subsequent washing of the silica beads with an ethanolic buffer set. The purified genomic DNA is eluted in a low ionic strength buffer at a concentration suitable for most downstream molecular biology applications.

Sera-Xtracta™ Genomic DNA Kit is supported by Cytiva's expertise in Sera-Mag bead technology. Automation scripts available, contact Scientific Support.

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    Sera-Xtracta Genomic DNA Kit
  • Cat. No.:
    29-4291-40
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Target molecule:
    DNA
  • Method / Format:
    Magnetic beads
  • Description:
    Sera-Xtracta Genomic DNA Kit
  • Sample type:
    Blood

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.