Order Entry
United Kingdom
ContactUsLinkComponent
VWR®, Wire basket with handle
VWR®, Wire basket with handle
Catalog # 216-2254
Supplier:  VWR Collection
undefined
VWR®, Wire basket with handle
Catalog # 216-2254
Supplier:  VWR Collection
Specifications
  • W×D×H:
    400×300×200 mm
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
This wire basket is made from rust-proof, electrolytically polished stainless steel.
  • Very good chemical and temperature resistance
  • Foldaway carrying handle with plastic hand grip
  • Robust design with two rods for base reinforcement
Suitable for transportation or sterilisation of laboratory goods.