Tetrahydrofuran, anhydrous ≥99.8%, AcroSeal® BHT-free, over molecular sieves
: Butylene oxide, THF, Diethylene oxide
:109-99-9
:
:C₄H₈O
:MFCD00005356
:66 °C
:–108.4 °C
:0.889
:72.11 g/mol
:–21°C
:203-726-8
:2056
:3 II
:12,09351
:Packaged under argon.
:GHS02
:GHS07
:GHS08
:Ambient
- Environmentally Preferable:
- Appearance:Clear Colorless liquid
- Assay (GC):≥99.8%
- Water content (Karl Fischer Titration):≤50 ppm
Frequently Bought Together