Hydrogen peroxide 34,5-36,5%, meets analytical specification of Ph. Nord.
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7722-84-1
:
Formula:H₂O₂
MDL Number:MFCD00011333
Boiling Pt:150.2
Melting Pt:–33 °C
Density:1.110 g/cm3 (20 °C)
Molecular Weight:34.01 g/mol
UN:2014
ADR:5.1 II
Merck Index:14,04798
:GHS07
:GHS05
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:34.5 - 36.5 %
- Non-volatile matter:Max. 0.05 %
- Residue on ignition:Max. 0.03 %
- Free acid (as H₂SO₄):Max. 0.02 %
- Heavy metals (as Pb):Max. 0.0002 %
Specification Test Results
Frequently Bought Together