Order Entry
United Kingdom
ContactUsLinkComponent
PCR clean-up reagent, Sera-Mag™ select
PCR clean-up reagent, Sera-Mag™ select
Supplier:  Cytiva
Certificates
undefined
PCR clean-up reagent, Sera-Mag™ select
Supplier:  Cytiva

Specifications

  • Application:
  • Method / Format:
  • Refine target molecule:
  • Sample type:
  • Target molecule:
  • Environmentally Preferable:

Specifications

Frequently Bought Together