Order Entry
United Kingdom
ContactUsLinkComponent
VWR® Traceable®, Temperature Data Loggers with TraceableGO™ Wireless Capability and Calibration
VWR® Traceable®, Temperature Data Loggers with TraceableGO™ Wireless Capability and Calibration
Supplier:  VWR Collection
Certificates
undefined
VWR® Traceable®, Temperature Data Loggers with TraceableGO™ Wireless Capability and Calibration
Supplier:  VWR Collection
Specifications
  • Catalog No:
  • 620-2734
  • 620-2736
  • 620-2735
  • Display:
  • Multiline LCD
  • Multiline LCD
  • Multiline LCD
  • Interface:
  • Bluetooth
  • Bluetooth
  • Bluetooth
  • Recording intervals:
  • 1 min - 12 h
  • 1 min - 12 h
  • 1 min - 12 h
  • Weight:
  • 18 g
  • 18 g
  • 18 g
  • W×D×H:
  • 89×20×51 mm
  • 89×20×51 mm
  • 89×20×51 mm
  • Data memory - No. of readings:
  • 65000
  • 65000
  • 65000
  • Batteries:
  • 2×AAA batteries (included)
  • 2×AAA batteries (included)
  • 2×AAA batteries (included)