Titriplex® IV solution c (DCTA Na₂) = 0,1 mol/l, Titripur® for metal titration, Supelco®
Catalog # 1.08447.1000
Supplier: Merck
CAS Number:
Specification Test Results
- Form:liquid
- Amount-of-substance concentration:0.0995 - 0.1005
- Measurement uncertainty:+/- 0.0003
- Traceability:NIST SRM
Specification Test Results
About this item
Synonyms: CDTA disodium salt
Molecular Weight:390.3 g/mol