Nitric acid : Dipicolinic acid (34 mM : 14 mM) in water eluent concentrate for IC (20x) for Metrosep C 4, Supelco®
: Merck
Nitric acid : Dipicolinic acid (34 mM : 14 mM) in water eluent concentrate for IC (20x) for Metrosep C 4, Supelco®
: Merck
: Nitric acid : Pyridine-2,6-dicarboxylic acid
:
- Environmentally Preferable:
Frequently Bought Together
Accessories for HPLC bottles, DURAN®
: DWK Life Sciences
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Crucibles, Porcelain
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
VWR®, Conductivity standard solutions
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection