Order Entry
United Kingdom
ContactUsLinkComponent
Ninhydrin reagent (includes buffer)
Ninhydrin reagent (includes buffer)
Catalog # HPLCN89001.
Supplier:  VWR Chemicals
CAS Number:  
undefined
Ninhydrin reagent (includes buffer)
Catalog # HPLCN89001.
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate
 :
 :178.14 g/mol
CAS Number:485-47-2
Formula:C₉H₆O₄
MDL Number:MFCD00003791
Boiling Pt:351
Merck Index:13,06582