Order Entry
United Kingdom
ContactUsLinkComponent
Hydrochloric acid diluted (60 ml dosed with 1 ml 37% HCl)
Hydrochloric acid diluted (60 ml dosed with 1 ml 37% HCl)
Catalog # 60HYD20252
Supplier:  VWR Chemicals
CAS Number:  
undefined
Hydrochloric acid diluted (60 ml dosed with 1 ml 37% HCl)
Catalog # 60HYD20252
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    HDPE bottle, wide mouth
  • Pk:
    96
  • Seq:
    2385
  • Environmentally Preferable:
  • Pk Info:
    60 ml dosed with 1 ml of 37% HCl

Specifications

About this item

Synonyms: Hydrochloric acid solution, Muriatic acid
 :
 :36.46 g/mol
UN:1789
 :GHS05
 :GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324
ADR:8 II