Pharmaceutical secondary standard, Lansoprazole, certified reference material, Supelco®
Catalog # PHR1390-1G
Supplier: Merck
CAS Number:
Specifications
- Concentration:Neat
- Pk:1 g
- Standards:Pharmaceutical secondary standard (CRM)
- CAS No.:103577-45-3
- Description:Lansoprazole
- Environmentally Preferable:
- Description:Lansoprazole
Specifications
About this item
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.