Pipette bulb, natural rubber, DURAN®
Catalog # SCOT292000205
Supplier: DWK Life Sciences
Specifications
- Capacity:2 ml
- Colour:Red
- Description:Pipette bulb, natural rubber, 14,3×42,4 mm (Ø×H)
- Environmentally Preferable:
- Pk:10
Specifications
About this item
Designed for removing or dispensing small quantities of liquids from Pasteur pipettes or droppers. The combination of the Pasteur pipette and rubber bulb can be used as a teat pipette.
- 5 mm opening
- Maximum operating temperature is 80 °C
The flexible rubber teat is suitable for use with Pasteur pipettes or droppers as a pipetting aid to dispense liquids, remove liquids, or add drops of reagents.
Packaging: 10 pieces supplied per pack.