Agmatine sulphate ≥97%, powder, Sigma-Aldrich®
Catalog # SIALA7127-1G
Supplier: Merck
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance (Color) :White to Off-White
- Appearance (Form) :Powder
- Infrared spectrum:≥97%
- Solubility (Turbidity)(50 mg/ml in H₂O):Clear to Slightly Hazy
- Solubility (Color) :Colorless to Faint Yellow
Specification Test Results
About this item
Synonyms: 1-Amino-4-guanidobutane Sulphate, 1-(4-Aminobutyl)guanidine sulphate
:
:228.27 g/mol
CAS Number:2482-00-0
Formula:C₅H₁₄N₄
MDL Number:MFCD00013109
Melting Pt:234…238 °C (lit.)
:M-Clarity™ quality level = MQ100