Eosin Y (yellowish) 1% alcoholic solution counterstain in H&E, for histology
1.17081.1000
: Merck
:
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
- Pk Info:Preparation Capacity 1000 - 1500 slides
- Suitability for microscopy (Tissue section):passes test
- Nuclei:dark blue to dark violet
- Cytoplasm:pink to red
- intercellular substances:pink to red
- Erythrocytes:yellow to orange
: Solvent Red 43, Field's Stain B, Eosin 225, 2,4,5,7-Tetrabromofluorescein disodium salt, Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoate, Disodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate), Tetrabromofluorescein disodium salt, 2′,4′,5′,7′-Tetrabromofluorescein disodium salt, Eosin Y disodium salt
:
:691.86 g/mol
:1170
:GHS02
:Ambient
:1
:17372-87-1
:C₂₀H₆Br₄Na₂O₅
:MFCD00005040
:3
:13,03634