Organic reference standard, Neotame, analytical standard, Supelco®
Catalog # 49777-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:165450-17-9
- Description:Neotame, ≥98,0% (HPLC)
- Environmentally Preferable:
- Formula:C₂₀H₃₀N₂O₅
- Description:Neotame
- Form:Neat
Specifications
About this item
Neotame is a white colored, dipeptide methyl ester, which is found to be a highly potent and non-nutritive sweetener or flavour enhancer and hence finds applications in variety of foods. It is manufactured from aspartame and 3,3-dimethylbutyraldehyde in a one-step chemical synthesis process.
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.