Order Entry
United Kingdom
ContactUsLinkComponent
Supelco® Organic Reference Standard, ʟ-Lysine Hydrochloride Solution
Supelco® Organic Reference Standard, ʟ-Lysine Hydrochloride Solution
Catalog # 44208-5ML-F
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, ʟ-Lysine Hydrochloride Solution
Catalog # 44208-5ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Standards:
    Analytical standard
  • CAS No.:
    10098-89-2
  • Description:
    ʟ-Lysine hydrochloride solution
  • Environmentally Preferable:
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H290
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00064564
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₆H₁₄N₂O₂.HCl
  • Molecular weight:
    182,65
  • Storage temperature:
    2…8 °C
  • Concentration:
    100 mm amino acid in 0,1 m HCL

Specifications

About this item

Caution: P234 - P390