Order Entry
United Kingdom
ContactUsLinkComponent
Chamber slides, C-Slide
Chamber slides, C-Slide
Catalog # NANOCS-050
Supplier:  NANOENTEK
undefined
Chamber slides, C-Slide
Catalog # NANOCS-050
Supplier:  NANOENTEK
Specifications
  • Recommended working volume:
    10 µl
  • Description:
    C-Slide cell counting chamber slides (100 tests)
  • Pk:
    50
Specifications
About this item
Cell counting chamber slides for use with automated cell counter (Countess and Countess II).
  • No need of coverslips
  • Eliminates the need of washing and reuse
  • Reduced exposures to the infectious samples and contamination
  • Guaranteed reproducibility and reliability
Made of sturdy and strong quartz grade optical plastic.