Supelco® PESTANAL® Organic Reference Standard, Terbutryn
Catalog # 45677-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:886-50-0
- Description:Terbutryn
- Seq:01
- Environmentally Preferable:
- Storage conditions:2...30 °C
- Hazard class description:H410
- Shipping conditions:STD
- MDL No.:MFCD00128025
- M-Clarity quality level:MQ100
- Formula:C₁₀H₁₉N₅S
- Molecular weight:241,36
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS07, GHS09
Caution: P273 - P391 - P501