Polyvinyl alcohol, M.W. 85000 - 124000 99+%
: Merck
: Poly(vinyl alcohol), PVOH, PVA, Mowiol®
:9002-89-5
:25213-24-5
:[-CH₂CH(OH)-]n
:MFCD00081922
:M-Clarity™ quality level = MQ200
- Environmentally Preferable:
- Appearance (Colour):White to Off-White
- Appearance (Form):Conforms to Requirements
- Infrared Spectrum:Conforms to Structure
- Miscellaneous Assay:99.3 - 100.0%
- Loss on Drying:≤5.0%
- Residue on Ignition:≤1.20%
- Viscosity, C=4%; Water:28.0 - 32.0 cps
- pH, C=4%; Water:5.5- 7.5
Frequently Bought Together