Vacuum concentrator, DNA SpeedVac system
531-1493
- Description:DNA SpeedVac system, 230 VAC, 50 Hz, 6 A
- Environmentally Preferable:
- W×D×H:290×64×31 mm
- Programs:3 selectable programs by application; can be modified and stored; auto and manual run selector
- Weight:48 kg
- Pk:1
Compact and integrated vacuum concentrator for sample safety and reproducible results.
- Compact, easy to use design
- Lid lock illuminates when lid is closed and locked
- External post trap neutralizes ammonium hydroxide vapors
- Data integrity and transfer ability to download data from a USB port
- Modes of operation: Auto, manual and pre-heat temperature
Designed for DNA and RNA applications, the compact and integrated Thermo Scientific™ Savant™ SpeedVac™ DNA130 vacuum concentrator supports the rapid and safe concentration and drying-down of samples, including nucleic acids, PCR preps and synthetic oligos.
For concentrating radioactive samples, an optional radioactive chemical trap kit can be used to minimize concerns in handling hazardous fumes and chemicals.
: Includes one RD36 rotor.