Lithium phenyl(2,4,6-trimethylbenzoyl)phosphinate (LAP) ≥95%, Sigma-Aldrich®
: Merck
: Lithium PTMB phosphinate, LAP, Phenyl(2,4,6-trimethylbenzoyl)phosphinic acid lithium salt, Lithium Phenyl(2,4,6-trimethylbenzoyl)phosphinate
:85073-19-4
:
:C₁₆H₁₆LiO₃P
:MFCD29472537
:294.21 g/mol
:Refrigerator
- Environmentally Preferable:
- Appearance (Color):White to Beige
- Appearance (Form):Powder or Crystals
- 1H NMR Spectrum:Conforms to Structure
- 31P NMR Spectrum:Conforms to Structure
- Carbon Content:62.0 - 68.6 %
- Purity (HPLC):≥ 95.0 %
Frequently Bought Together