Order Entry
United Kingdom
ContactUsLinkComponent
BCECF AM ester 1 mg/mL in DMSO fluorescent pH indicator, Sigma-Aldrich®
BCECF AM ester 1 mg/mL in DMSO fluorescent pH indicator, Sigma-Aldrich®
Catalog # B8806-1MG
CAS Number:  
BCECF AM ester 1 mg/mL in DMSO fluorescent pH indicator, Sigma-Aldrich®
Catalog # B8806-1MG
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 2′,7′-Bis(2-carboxyethyl)-5(6′)-carboxyfluorescein acetoxymethyl ester, Spiro(isobenzofuran-1(3H),9'-(9H)xanthene)-2',7'-dipropanoic acid, BCECF AM
 :
 :808.69 g/mol
Storage Temperature:Freezer
CAS Number:117464-70-7
Formula:C₄₂H₄₀O₂₁
MDL Number:MFCD00036969
 :M-Clarity™ quality level = MQ200