Order Entry
United Kingdom
ContactUsLinkComponent
Microscope lens cleaning kit
Microscope lens cleaning kit
Catalog # MEDL2480.2075
Supplier:  MEDLINE SCIENTIFIC
undefined
Microscope lens cleaning kit
Catalog # MEDL2480.2075
Supplier:  MEDLINE SCIENTIFIC
Specifications
  • Description:
    Microscope cleaning kit
  • Environmentally Preferable:
  • Pk:
    1 SET
Specifications
About this item
This cleaning kit ensure the microscope cleanliness for optimal viewing. It contains cleaning wipes, solution and air brush.
  • Cleaning tissue for removal of oil from immersion objectives
  • Air brush for routine dust removal
  • Lens cleaning fluid and a cleaning cloth for cleaning eyepieces and glass components
Delivery: Kit includes lens cleaning tissues (20 pcs), lens cleaning solution (75 ml) and air brush.