Stearyl Stearate
Catalog # REST31860
Supplier: Restek
CAS Number:
Specifications
- Concentration:Neat
- Pk:1 Ampoul
- CAS No.:2778-96-3
- Description:Stearyl stearate
- Environmentally Preferable:
- Formula:C₃₆H₇₂O₂
- Description:Stearyl stearate
- Pk Info:100 mg/ampoule
Specifications
About this item
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.