Order Entry
United Kingdom
ContactUsLinkComponent
Mineral oil light USP, NF, Ph. Eur., BP, pharma grade
Mineral oil light USP, NF, Ph. Eur., BP, pharma grade
Catalog # APPC146257.1211
Supplier:  PanReac AppliChem
CAS Number:  
Mineral oil light USP, NF, Ph. Eur., BP, pharma grade
Catalog # APPC146257.1211
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pk:
    1.000 mL
  • Seq:
    2865
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Identity According to Pharmacopoeias:
    Passes Test
  • Density 20/20:
    0.810 - 0.875
  • Density 25/25:
    0.818 - 0.880
  • Viscosity Kinematic (39.9 - 40.1 °C):
    3.0 - 33.5 mm²/s
  • Viscosity Dynamic (19.9 - 20.1 °C):
    25 - 80 mPas
  • Acidity or Alkalinity:
    Passes Test
  • Darkened Substances by H₂SO₄:
    Passes Test
  • Sulphur Compounds:
    Passes Test
  • Solid Paraffin:
    Passes Test
  • Residual Solvents (Ph.Eur./USP):
    Passes Test
  • Polycyclic Aromatic Hydrocarbons:
    Passes Test

Specification Test Results

About this item

 :
EINECS:232-455-8
 :GHS08
CAS Number:8042-47-5
MDL Number:MFCD00131611
Boiling Pt:300...450 °C
Density:0.845 g/cm³
Flash Pt:>180 °C