Order Entry
United Kingdom
ContactUsLinkComponent
Silicone DC 710 fluid
Silicone DC 710 fluid
Catalog # SERA35149.04
Supplier:  Serva
CAS Number:  
Silicone DC 710 fluid
Catalog # SERA35149.04
Supplier:  Serva
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Description:
    DC 710
  • Seq:
    07
  • Environmentally Preferable:
  • Flash point:
    302 °C
  • Viscosity:
    500 mm²/s (25 °C)

Specifications

About this item

  • Retains its properties after continuous use at temperatures up to 260°C in closed systems
  • Shows very low volatility even at elevated temperatures
  • Highly resistant to oxidation and gumming