(1R,2R)-(+)-1,2-Cyclohexanediamine-L-tartrate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: (1R,2R)-Cyclohexane-1,2-diamine (2R,3R)-2,3-dihydroxysuccinate, (1R)-trans-1,2-Diaminocyclohexane L-Tartrate
CAS Number:39961-95-0
:
Formula:C₆H₁₄N₂
MDL Number:MFCD00191979
Molecular Weight:264.28 g/mol
Melting Pt:273 °C (dec.)
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:≥97,5
Specification Test Results
Related Information
Frequently Bought Together