Labels, pressure-sensitive
Supplier: Thermo Fisher Scientific
Certificates
About this item
These Nalgene™ polyolefin pressure-sensitive labels are used with photocopiers, laser printers or inkjet printers. Accepts a variety of inks including copier, marker, ballpoint, gels, and technical markers (pretest specific inks for suitability prior to use), does not accept pencil.
- Rated to −29 °C
- Resistant to ultraviolet light, mildew, chemicals and dirt and won't discolor, crack or tear
- Excellent for labeling microscopic slides - adhesive backing sticks well to any plastic, glass or metal surface
- Chemically compatible with most lab reagents although solvents may dissolve adhesive
- Autoclavable
Formatted for easy setup in word processing programs; instructions included in each pack.
Specifications
- Catalog No:
- 817-0082
- 817-0083
- 817-0084
- 817-0085
- 817-0081
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement