Order Entry
United Kingdom
ContactUsLinkComponent
Hexamethylenetetramine
Hexamethylenetetramine
Catalog # HACH2603999
Supplier:  Hach
CAS Number:  
undefined
Hexamethylenetetramine
Catalog # HACH2603999
Supplier:  Hach
CAS Number:  

Specifications

  • Pk:
    100
  • Seq:
    0100
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Hexamethylenetetramine, Hexamine, Urotropine, Formin
 :
 :140.19 g/mol
Flash Pt:250
EINECS:202-905-8
UN:1328
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:100-97-0
Formula:C₆H₁₂N₄
MDL Number:MFCD00006895
Melting Pt:280
Density:1.33 g/cm³ (20 °C)
ADR:4.1 III
Merck Index:13,05994