Pyridine, Sigma-Aldrich®
: Merck
: Azabenzene, Azine
:110-86-1
:
:C₅H₅N
:MFCD00011732
:115
:–44 °C
:0.983 g/cm³ (20 °C)
:79.1 g/mol
:17
:203-809-9
:1282
:3 II
:13,08060
:M-Clarity™ quality level = MQ300
:GHS02
:GHS07
:Ambient
- Environmentally Preferable:
Frequently Bought Together