Isonicotinic acid
Catalog # PROOMM0553.03
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
- Environmentally Preferable:
Specifications
About this item
Synonyms: Pyridine-4-carboxylic acid, 4-Pyridinecarboxylic acid, 4-Picolinic acid
:
:123.11 g/mol
EINECS:200-228-2
:GHS07
Storage Temperature:Ambient
CAS Number:55-22-1
Formula:C₆H₅NO₂
MDL Number:MFCD00006429
Boiling Pt:396
Melting Pt:315…319 °C
Merck Index:13,05206