Zinc chloride, anhydrous 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Zinc dichloride, Dichlorozinc, Zinc (II) chloride
CAS Number:7646-85-7
:
Formula:ZnCl₂
MDL Number:MFCD00011295
Boiling Pt:732 °C
Melting Pt:290 °C
Density:2.91
Molecular Weight:136.3 g/mol
Storage Temperature:Ambient
EINECS:231-592-0
ADR:8 III
Merck Index:14,10132
UN:2331
:GHS05
:GHS07
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together