Order Entry
United Kingdom
ContactUsLinkComponent
Copper(II) oxide 99+%
Copper(II) oxide 99+%
Certificates
undefined
Copper(II) oxide 99+%
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Black Powder
  • Titration Complexometric:
    ≥99.0 %
  • Chloride (Cl):
    ≤50 ppm
  • Carbon (C):
    ≤500 ppm
  • Iron (Fe):
    ≤500 ppm
  • Total sulfur (as SO4):
    ≤0.02 %
  • Free alkali:
    Passes test
  • Insoluble matter:
    ≤0.02 % (in dilute HCl)
  • Nitrogen compounds (as N):
    ≤0.002 %

Specification Test Results

Related Information

Frequently Bought Together