Silver(II) fluoride
Supplier: FLUOROCHEM
Certificates
About this item
Synonyms: Silver difluoride, Silver(II)fluoride
CAS Number:7783-95-1
:
Formula:AgF₂
MDL Number:MFCD00003411
Density:4.57…4.78 g/cm³ (18 °C)
Molecular Weight:145.86 g/mol
Melting Pt:690
ADR:8 II
Merck Index:12,08656
EINECS:232-037-5
UN:3084
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together