Methyltrimethoxysilane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Trimethoxy(methyl)silane
CAS Number:1185-55-3
:
Formula:C₄H₁₂O₃Si
MDL Number:MFCD00008342
Boiling Pt:102
Melting Pt:11
Density:0.955 g/cm³ (20 °C)
Molecular Weight:136.22 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:214-685-0
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together