Beryllium oxide ≥99% (metals basis)
Catalog # 10001.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
About this item
:
:25.01 g/mol
EINECS:215-133-1
UN:1566
:GHS06
:GHS08
:GHS07
Storage Temperature:Ambient
CAS Number:1304-56-9
Formula:BeO
MDL Number:MFCD00003457
Boiling Pt:4300 °C
Melting Pt:2575 °C
Density:3.01
ADR:6.1 II
Merck Index:14,1172