Magnesium iodide
Catalog # MOLE14708297-5G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
About this item
:
:278.11 g/mol
EINECS:233-825-1
CAS Number:10377-58-9
Formula:MgI₂
MDL Number:MFCD00049485
Melting Pt:637
Density:3.89 g/cm³ (20 °C)
Merck Index:12,05707