Magnesium hydroxide carbonate light (∼40,0 - 45,0% MgO) for analysis
: Magnesium carbonate basic, Magnesium carbonate hydroxide, Magnesium carbonate hydrated basic
:39409-82-0
:MFCD00149783
:MgCO₃ (linear)
:MFCD07371358
:3600
:2.16 g/cm³
:84.31 g/mol
:Ambient
:235-192-7
- Environmentally Preferable:
- Appearance:White Powder
- Assay:40.0 to 45.0 % (MgO)
- Chloride (Cl):≤700 ppm
- Sulfate (SO4):≤3000 ppm
- Arsenic (As):≤2 ppm
- Calcium (Ca):≤7500 ppm
- Iron (Fe):≤400 ppm
- Heavy metals (as Pb):≤20 ppm
- Soluble matter:≤1 % (in water)
- Insoluble matter:≤0.05 % (in acetic acid)
Frequently Bought Together