Calcium carbonate ≥99.5% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Marble
CAS Number:471-34-1
:
Formula:CaCO₃
MDL Number:MFCD00010906
Density:2.930
Molecular Weight:100.09 g/mol
Melting Pt:800 °C (dec.)
Merck Index:14,1657
Storage Temperature:Ambient
EINECS:207-439-9
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together