≥97%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate
CAS Number:7758-11-4
:
Formula:K₂HPO₄
MDL Number:MFCD00011383
Density:2.3 g/cm³ (20 °C)
Molecular Weight:174.18 g/mol
Melting Pt:340
Merck Index:13,07743
Storage Temperature:Ambient
EINECS:231-834-5
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:Min. 97 %
- Heavy metals (as Pb):Max. 50 ppm
- Cl (Chloride):Max. 0.02 %
- SO4 (Sulphate):Max. 0.05 %
- Fe (Iron):Max. 50 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 2,5L Plastic bottle for solids DIN 80 (ZPACK13252500) 20 L Plastic bucket for solids UN Z (ZPACK13247020)
Packaging Information
26930_DI-POTASSIUM HYDROGEN PHOSPHATE_BSE_REV0
BSE/TSE Certificates
spec_VWRC26930.260 spec_VWRC26930.260 spec_VWRC26930.293 spec_VWRC26930.293 spec_VWRC26930.362
Product specification
More
Frequently Bought Together