Potassium hydroxide, pellets, EMSURE® for analysis, Supelco®
: Merck
: Potash caustic, Caustic potash
:1310-58-3
:
:KOH
:MFCD00003553
:1320
:360
:2.04 g/cm³ (20 °C)
:56.11 g/mol
:Ambient
:215-181-3
:1813
:8 II
:12,07806
:M-Clarity™ quality level = MQ300
:01-2119487136-33
:GHS05
:GHS07
- Environmentally Preferable:
- Solubility:1130 g/l (20 °C)
- Melting Point:360 °C
- Molar Mass:56.11 g/mol
- Boiling Point:1320 °C
- Vapor Pressure:(20 °C)
- Density:2.04 g/cm3 (20 °C)
- pH:14 (56 g/l, H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Frequently Bought Together