Fluorescein disodium salt, TECHNICAL
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: 3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, disodium salt, Acid Yellow 73, Yellow No. 8, Fluorescein sodium salt, 2-(6-hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid disodium salt, Disodium 2-(3-oxo-6-oxidoxanthen-9-yl)benzoate
CAS Number:518-47-8
:
Formula:C₂₀H₁₀Na₂O₅
MDL Number:MFCD00167039
Density:1.601 g/cm³ (20 °C)
Molecular Weight:376.28 g/mol
Melting Pt:> 300 °C
Merck Index:13,04185
Storage Temperature:Ambient
EINECS:208-253-0
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appareance:Passes test
- IR Spectrum:Passes test
- UV Spectrum:Passes test
- Absorption max. (buffer pH 8,0):490 - 492 nm
- Spec.absorptivity(Lm;0,005g/l;pH8,0;dry):Min. 1200
- Assay:Min. 80%
- Loss on drying (110°C) at manufacturing:Max. 10 %
Specification Test Results
Related Information
Certificates
Search by Lot Number20 L Plastic bucket for solids UN Z (ZPACK13247020)
Packaging Information
spec_VWRC260983T spec_VWRC260983T spec_VWRC26098CF spec_VWRC26098CF
Product specification
Frequently Bought Together