Decamethylcyclopentasiloxane ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Cyclopentasiloxane, 2,2,4,4,6,6,8,8,10,10-Decamethyl-1,3,5,7,9,2,4,6,8,10-pentaoxapentasilecane, Dow Corning 245® fluid, Volasil 245, Silicone fluid DC 245
CAS Number:541-02-6
:
Formula:C₁₀H₃₀O₅Si₅
MDL Number:MFCD00046966
Boiling Pt:210 °C
Melting Pt:–44 °C
Density:0.959
Molecular Weight:370.77 g/mol
Flash Pt:76 °C (169 °F)
Merck Index:12,02905
Storage Temperature:Ambient
EINECS:208-764-9
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together